Learn More
Glas Col LLC Rotator
SDP
Catalog No. NC9965209
Encompass_Preferred
Mixing action from a gentle slosh to rapid agitation. Fixed 20 degree angle, furnished with alumninum disk head with clips for 5-10mm and 12-14mm tubes. Variable speed,2-80 rpm, 1.5A. 50/60Hz, 120V
Catalog No. NC9965209 Supplier Glas Col LLC Supplier No. 099A MR1512
Mixing action from a gentle slosh to rapid agitation. Fixed 20 degree angle, furnished with alumninum disk head with clips for 5-10mm and 12-14mm tubes. Variable speed,2-80 rpm, 1.5A. 50/60Hz, 120V