Learn More
World Precision Instrument Mini Dissecting Scissors, 8.5cm, Curved, Sharp tips
SDP
Catalog No. 50380014
Encompass
Our Mini Dissecting Scissors are similar to regular dissecting scissors, but they are shorter and have more delicate tips, making them ideal for fine dissection work in tight spaces. They are commonly used in microsurgery, like ophthalmic surgery. These scissors are 8.5 cm (3.3") long and have straight or curved blades with blunt or sharp tips and fine blades. Our dissecting scissors are for general use in dissection, preparation of tissue samples, and small animal surgeries. Dissecting scissors have thinner (and often longer) blades than operating scissors. They are designed for cutting and probing in tight spaces. These are available in surgical grade stainless steel. Our Dissecting Scissors are designed to withstand the high heat and chemical exposure of the sterilization process. They may be autoclaved. To guarantee a long life for your surgical instruments, be sure to clean them thoroughly after each use.
Catalog No. 50-380-014 Supplier World Precision Instrument Supplier No. 503666
Our Mini Dissecting Scissors are similar to regular dissecting scissors, but they are shorter and have more delicate tips, making them ideal for fine dissection work in tight spaces. They are commonly used in microsurgery, like ophthalmic surgery. These scissors are 8.5 cm (3.3") long and have straight or curved blades with blunt or sharp tips and fine blades. Our dissecting scissors are for general use in dissection, preparation of tissue samples, and small animal surgeries. Dissecting scissors have thinner (and often longer) blades than operating scissors. They are designed for cutting and probing in tight spaces. These are available in surgical grade stainless steel. Our Dissecting Scissors are designed to withstand the high heat and chemical exposure of the sterilization process. They may be autoclaved. To guarantee a long life for your surgical instruments, be sure to clean them thoroughly after each use.