Learn More
Mopec™ Mini Section Dryer Timer
SDP
Catalog No. 22445555 Shop All Mopec Products
Change view
Click to view available options
Product Type:
Microscope Slide Dryer
Catalog No. Product Type
22-445-555 Microscope Slide Dryer
1 options
Catalog No. 22-445-555 Supplier Mopec™ Supplier No. BK700T
Add to Cart
Add to Cart
The Mini Section Dryer is designed to efficiently and rapidly blow temperature-controlled air through the base of the instrument to dry specimens without the risk of overheating. Through this process, drying times are dramatically reduced by 75% compared to conventional drying ovens.
  • Fan-assisted heating dramatically reduces drying time
  • Designed to take up minimal bench-top space
  • Digital temperature control
  • Rapid heating system ensures system is ready to use in less time
  • Anti-microbial coating to help prevent bacteria and virus growth
  • Timer Unit allows monitoring for up to four slides and ensures each rack has the correct amount of time in the dryer
Specifications
Product Type Microscope Slide Dryer
For Use With (Equipment) Mini Section Dryer