Learn More
Electron Microscopy Sciences Mini SEM Clamp,
SDP
Catalog No. 501927142
Encompass_Preferred
Mini SEM Clamp on a standard 12.7 mm ( 1/2") pin stub. Suitable for holding flat or thin specimens such as silicon chips, foil, wires, and small tubes. Material: Vacuum-grade aluminum with brass screw.
Pin length: 12.7 mm
Catalog No. 50-192-7142 Supplier Electron Microscopy Sciences Supplier No. 7595401
Mini SEM Clamp on a standard 12.7 mm ( 1/2") pin stub. Suitable for holding flat or thin specimens such as silicon chips, foil, wires, and small tubes. Material: Vacuum-grade aluminum with brass screw.
Pin length: 12.7 mm