Learn More
Bellco Glass, Inc. Mini-Shaker Base Only, 115V
SDP
Catalog No. 501946139
Encompass_Preferred
The Bellco Benchtop Mini Orbital Shakers are ideal for both rigid polystyrene and flexible polyvinyl 96-well micro tube plates commonly used in cell culture, virology, immunology and serology. They are engineered to operate between thespeed range of 50 to 1550 rpm and have a 100 second timer for reproducing mixing times or can be used in continuous operation. The true orbital motion results in increased mixture homogeneity compared with oscillating or vibrating shakers. The cast iron ballast eliminates walking of unit and handles allow easy movement within the laboratory.. MADE IN THE USA
Catalog No. 50-194-6139 Supplier Bellco Glass, Inc. Supplier No. 774408115
The Bellco Benchtop Mini Orbital Shakers are ideal for both rigid polystyrene and flexible polyvinyl 96-well micro tube plates commonly used in cell culture, virology, immunology and serology. They are engineered to operate between thespeed range of 50 to 1550 rpm and have a 100 second timer for reproducing mixing times or can be used in continuous operation. The true orbital motion results in increased mixture homogeneity compared with oscillating or vibrating shakers. The cast iron ballast eliminates walking of unit and handles allow easy movement within the laboratory.. MADE IN THE USA