Learn More
Bellco Glass, Inc. Mini-Shaker Tray,32 Pos
SDP
Catalog No. 501946142
Encompass_Preferred
Material: .125" alumDimensions: 16 1/4" w x 10 5/8" d x 3 1/8" tPositions: 32. MADE IN THE USA
Catalog No. 50-194-6142 Supplier Bellco Glass, Inc. Supplier No. 774408240
Material: .125" alumDimensions: 16 1/4" w x 10 5/8" d x 3 1/8" tPositions: 32. MADE IN THE USA