Learn More
Sigma Aldrich Fine Chemicals Biosciences Mini-sieve micro sieve set
Catalog No. 502900532
Encompass
Hand held sieve for quick field or lab particle size sorting. Supplied with four exchangeable screen-holding segments a catch pan a cover and four screen-retaining rings. Each storing segment 51mm x 29mm (2 x 1 1/8) can contain approximately 60cc. System comes with one each 25 35 45 60 80 120 170 and 230 standard mesh screens.Replacement screen inserts available (Z675423)Overall dimensions 164mm x 73mm (6 7/16 x 2 7/8)Polypropylene segments and collection pan assembles/disassembles for easy sample insertion and retrieval.Sieving specifications rapidly altered by exchanging the four detachable phosphor-bronze mesh screens.
Catalog No. 50-290-0532 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z6754151SET
Hand held sieve for quick field or lab particle size sorting. Supplied with four exchangeable screen-holding segments a catch pan a cover and four screen-retaining rings. Each storing segment 51mm x 29mm (2 x 1 1/8) can contain approximately 60cc. System comes with one each 25 35 45 60 80 120 170 and 230 standard mesh screens.Replacement screen inserts available (Z675423)Overall dimensions 164mm x 73mm (6 7/16 x 2 7/8)Polypropylene segments and collection pan assembles/disassembles for easy sample insertion and retrieval.Sieving specifications rapidly altered by exchanging the four detachable phosphor-bronze mesh screens.