Learn More
LUNA NANOTECH INC Mini Silica Spin Columns, 500 Columns
Supplier:  LUNA NANOTECH INC USP004500N
Encompass_Preferred
PuroSPIN™ MINI Spin Columns for DNA / RNA Extraction and Purification, 500 columns
Have you ever ran out of columns that come with your favorite nucleic acid extraction kit but still had a lot of left-over buffer reagents remaining? Instead of buying a new expensive kit you can use PuroSPIN™ columns with the left-over buffers as a cost-effective alternative. Simply use PuroSPIN™ columns while following the manufacturer-specific protocols.
 PuroSPIN™ MINI silica spin columns bind both DNA and RNA
200 μg genomic DNA and RNA binding capacity
Compatible as cost-effective replacement columns for nucleic acid extraction kits from Qiagen, ThermoFisher, Bio-Rad, Promega and other manufacturers
Each item includes both the spin column and a 2 mL sample collection tube
Extraction of DNA from bacteria, viruses, yeast, cultured cells, blood, urine, buccal swabs, animal tissue
Catalog No. 50-256-6907