Learn More
MSC Value Collection Mini Sine Bar, 5 in., V-Groove, 1 ea.
Catalog No. 5000511121
Encompass
The 5-in. mini sine bar features a v-groove design, providing a stable platform for precision angle setting and layout applications in metalworking environments.
  • Features a 5-in. length
  • Designed with a v-groove for secure workpiece alignment
  • Engineered for precision layout and angle measurement
Catalog No. 50-005-11121 Supplier MSC Supplier No. 03293578
The 5-in. mini sine bar features a v-groove design, providing a stable platform for precision angle setting and layout applications in metalworking environments.
  • Features a 5-in. length
  • Designed with a v-groove for secure workpiece alignment
  • Engineered for precision layout and angle measurement