Learn More
Mopec Mini Slide Drying Hotplate
SDP
Catalog No. 22445723 Shop All Mopec Products
Change view
Click to view available options
Dimensions (L x W x H):
145 x 145 x 65 mm
Catalog No. Dimensions (L x W x H)
22-445-723 145 x 145 x 65 mm
1 options
Catalog No. 22-445-723 Supplier Mopec Supplier No. BK402
Add to Cart
Edge
Add to Cart
Mini Slide Drying Hotplate is designed to quickly and efficiently remove wrinkles from tissue sections while taking up minimal bench-top space. The rapid heating system ensures the hotplate is ready to use in less time.
  • Designed to take up minimal bench-top space
  • Rapid heating system ensures system is ready to use in less time
  • Anti-microbial coating to help prevent bacteria and virus growth
Specifications
Product Type Mini Hotplate
Material Stainless Steel
Media Capacity 1
Temperature Range (Metric) Ambient to 80°C (at 20°C)
Dimensions (L x W x H) 145 x 145 x 65 mm
Voltage 110/230 V