Learn More
BURKLE INC Mini solvent pump, 0.48 gal. (1,8 l)/min, connection GL45
Catalog No. 5000025719
Encompass_Preferred
The Mini solvent pump is the ideal solution for safely dispensing small quantities of solvents, chemicals, and highly flammable liquids from bottles, canisters, and containers up to approx. 10 L. Using a positive pressure system, only the PTFE hose, pump head, and outlet touch the liquid, ensuring superior chemical resistance and purity. All wetted parts are made from high-quality PTFE and stainless steel, perfect for acetone, isopropanol, ethanol, methanol, and similar media. A built-in pressure relief valve protects against heat-related overpressure. The adjustable PTFE hose allows complete emptying, even from curved-bottom containers. Standard GL 45 thread fits adapters for S 40, GL 38, or GL 32. The self-closing, drip-free outlet enables accurate filling of even small vessels or test tubes, with optional hose for remote filling. Includes grounding cable for safe use in explosive atmospheres. Compact, tilt-resistant, and durable for clean, efficient, and long-lasting operation.
Catalog No. 50-000-25719 Supplier BURKLE INC Supplier No. 56034000
The Mini solvent pump is the ideal solution for safely dispensing small quantities of solvents, chemicals, and highly flammable liquids from bottles, canisters, and containers up to approx. 10 L. Using a positive pressure system, only the PTFE hose, pump head, and outlet touch the liquid, ensuring superior chemical resistance and purity. All wetted parts are made from high-quality PTFE and stainless steel, perfect for acetone, isopropanol, ethanol, methanol, and similar media. A built-in pressure relief valve protects against heat-related overpressure. The adjustable PTFE hose allows complete emptying, even from curved-bottom containers. Standard GL 45 thread fits adapters for S 40, GL 38, or GL 32. The self-closing, drip-free outlet enables accurate filling of even small vessels or test tubes, with optional hose for remote filling. Includes grounding cable for safe use in explosive atmospheres. Compact, tilt-resistant, and durable for clean, efficient, and long-lasting operation.