Learn More
Med Vet International Mini Spay Hook, Stainless Steel, 6 3/4", J0112M, J0112M
Catalog No. 5000584040
Encompass_Preferred
The first spay hook small enough for kittens and puppies. The increasing popularity of spaying at younger ages and small sizes makes the demand for smaller equipment. The spay hook has no ball and a narrow hook width of 3mm versus standard 5mm and a shorter length of 6 1/2in.
Catalog No. 50-005-84040 Supplier Med Vet International Supplier No. JOR112M
The first spay hook small enough for kittens and puppies. The increasing popularity of spaying at younger ages and small sizes makes the demand for smaller equipment. The spay hook has no ball and a narrow hook width of 3mm versus standard 5mm and a shorter length of 6 1/2in.