Learn More
Hanna Pharmaceutical Supply Co., Inc. MINI-SPIKE Dispensing Pin with SAFSITE Valve
Catalog No. NC1709765
Encompass_Preferred
Automatic two-way valve for aspiration or injection of medication from inverted multi-dose vials Utilizes a bacterial retentive air venting filter. Luer slip connector. 50/CS
Please note: This item is nonrefundable, non-returnable and non-cancelable once order is placed.
This item is not returnable. View return policy
Catalog No. NC1709765 Supplier Hanna Pharmaceutical Supply Co., Inc. Supplier No. BBR412000
Automatic two-way valve for aspiration or injection of medication from inverted multi-dose vials Utilizes a bacterial retentive air venting filter. Luer slip connector. 50/CS
Please note: This item is nonrefundable, non-returnable and non-cancelable once order is placed.