Learn More
Med Vet International Mini-Spike Withdrawal and Injection Spike, J1618, Each
Catalog No. 5000583065
Encompass_Preferred
The Mini-Spike withdrawal and injection spike for multi-dose containers is a simple device that allows hygienic syringe withdrawal from multi-dose liquid medication bottles. N User Advantages • Integrated, bacteria-proof 0.45 µm aeration filter • Green snap-on lid to protect luer lock connection • Additional protection against contamination • No manual pressure equalization required • No air bubbles when filling • Fill in rapid succession • Easy dosage measurement • Luer slip or luer lock syringes J1618 Mini-Spike. For multiple-withdrawal containers in which medication is already in liquid form.
Catalog No. 50-005-83065 Supplier Med Vet International Supplier No. JOR1618
The Mini-Spike withdrawal and injection spike for multi-dose containers is a simple device that allows hygienic syringe withdrawal from multi-dose liquid medication bottles. N User Advantages • Integrated, bacteria-proof 0.45 µm aeration filter • Green snap-on lid to protect luer lock connection • Additional protection against contamination • No manual pressure equalization required • No air bubbles when filling • Fill in rapid succession • Easy dosage measurement • Luer slip or luer lock syringes J1618 Mini-Spike. For multiple-withdrawal containers in which medication is already in liquid form.