Learn More
Epoch Life Science Inc DNA/RNA 250 MinispinCol W/Lid
SDP
Catalog No. NC0066803
Encompass_Preferred
Bed volume of 800ul, binding capacity of 30-40ug allow them to be directly plugged into your existing protocol.Sold with 2 ml collection tubes.Fits most vacuum manifold.
Catalog No. NC0066803 Supplier Epoch Life Science Inc Supplier No. 1920250
Bed volume of 800ul, binding capacity of 30-40ug allow them to be directly plugged into your existing protocol.Sold with 2 ml collection tubes.Fits most vacuum manifold.