Learn More
Epoch Life Science Inc EconoSpin® All-In-One DNA/RNA Mini Spin Columns w/o lid (50 columns with collection tubes)
SDP
Catalog No. NC1057003
Encompass_Preferred
Silica membrane mini spin columns with collection tubes for gel PCR & plasmid purification. Sample volume: 850 ul. Binding capacity: 45 ug plasmid DNA. Minimum elution volume: 20 ul. Compatible with third party buffer sets. This item does not include a lid. 18 month shelf life. Used by labs around the world.
Catalog No. NC1057003 Supplier Epoch Life Science Inc Supplier No. 1910050
Silica membrane mini spin columns with collection tubes for gel PCR & plasmid purification. Sample volume: 850 ul. Binding capacity: 45 ug plasmid DNA. Minimum elution volume: 20 ul. Compatible with third party buffer sets. This item does not include a lid. 18 month shelf life. Used by labs around the world.