Learn More
Epoch Life Science Inc EconoSpin® Silica DNA Midi Columns (50/PK)
SDP
Catalog No. NC0892479
Encompass_Preferred
Silica membrane columns for large scale non-transfection grade plasmid DNA extraction. 250 ug plasmid DNA binding capacity. 6.5 ml bed volume. Fits into most 15ml centrifuge/culture tubes. Requires proper centrifuge and rotor. Less expensive and faster alternative to anion-exchange columns. For DNA used for cell transfection please try our anion exchange columns (1970050) instead.
Catalog No. NC0892479 Supplier Epoch Life Science Inc Supplier No. 2030050
Silica membrane columns for large scale non-transfection grade plasmid DNA extraction. 250 ug plasmid DNA binding capacity. 6.5 ml bed volume. Fits into most 15ml centrifuge/culture tubes. Requires proper centrifuge and rotor. Less expensive and faster alternative to anion-exchange columns. For DNA used for cell transfection please try our anion exchange columns (1970050) instead.