Learn More
Hoefer, Inc Hoefer™ SUB6 Mini Submarine Gel Electrophoresis Unit
SDP
Supplier:  Hoefer, Inc SUB6
Encompass_Preferred
Ideal for routine preparatory and analytical electrophoresis techniquesFeatures and Benefits:Color-coded and height adjustable combs—easily identify comb thickness at a glance and control well depthColored loading strips—for easy well detection when loading Compact tank—reduces the buffer volume required to cover the gel, providing greater control over the voltage gradient and run-timeUV-transparent running tray—allows the user to image the gel without risk of damage due to handlingSide handles—for safe and easy transportation around the laboratoryIncludes:Buffer Chamber; Safety Lid; High Voltage Leads; Running Tray with Silicone Gasket; 1 mm thick, 8-well combs, 2 pcs; Colored Loading StripsSpecifications:Gel Dimensions (w x l): 6 x 7.5 cmMaximum Buffer Volume: 325 mlMaximum Power Settings: 300 V, 200 mA, 60 WMaximum Sample Capacity: 32Unit Dimensions (w x l x h): 13 x 24 x 6.5 cmSafety Certifications: EN61010-1, CE
Catalog No. NC2741101