Learn More
Biotium Mini Syringe Filtration Kit (for 5 samples)
Supplier:  Biotium 22026
Encompass_Preferred
Mini syringe filters are convenient for sterile filtration of small volumes (1 mL or less) with only 10 uL retention volume. Millex-GV 0.22 um hydrophilic PVDF membrane in a 4 mm filter. Kit includes sterile filters, 1 cc syringes, and screw cap vials, 5 each.
Catalog No. 50-196-4401