Learn More
Cole-Parmer Digi-Sense Miniature Type-K Thermocouple Male Connector, 2 Pin, 5Pk
Catalog No. 501961549 Shop All Cole-Parmer Products
Encompass_Preferred
Miniature connectors are suited for small wires and mineral insulated cables. Glass-filled nylon construction with hollow pins and molded barriers prevent short circuit. Secure thermocouple wire into positive/negative terminals with screws. Each connector comes in ANSI color coded material: type J-black, type K-yellow, and type T-blue.
Catalog No. 50-196-1549 Supplier Cole-Parmer Supplier No. 1852690
Miniature connectors are suited for small wires and mineral insulated cables. Glass-filled nylon construction with hollow pins and molded barriers prevent short circuit. Secure thermocouple wire into positive/negative terminals with screws. Each connector comes in ANSI color coded material: type J-black, type K-yellow, and type T-blue.