Learn More
Research Products International Corp Mini-Time Controller
SDP
Catalog No. 50212768
Encompass
Versatile and economical controller may be programmed to turn equipment on and off at any minute of any day. A fail-safe memory permits programming 14 different switchings per day. Random function automatically turns the controller on and off at random intervals. Schedule an entire week with the press of a few buttons. Switches located on the front panel control the entire operation. The Outlet Controller will turn equipment on and off in intervals as short as 1 minute or as long as 168 hours. Turn an instrument on today and off a week later. The AM/PM day programs allow repeating the same cycle every day of the week, weekdays only, weekends only, or any selected days.
To assure accuracy an individually serial-numbered Traceable Certificate is provided from our ISO 17025 calibration laboratory accredited by A2LA. It indicates traceability to standards provided by NIST (National Institute of Standards and Technology).
Catalog No. 50-212-768 Supplier Research Products International Corp Supplier No. 810030
Versatile and economical controller may be programmed to turn equipment on and off at any minute of any day. A fail-safe memory permits programming 14 different switchings per day. Random function automatically turns the controller on and off at random intervals. Schedule an entire week with the press of a few buttons. Switches located on the front panel control the entire operation. The Outlet Controller will turn equipment on and off in intervals as short as 1 minute or as long as 168 hours. Turn an instrument on today and off a week later. The AM/PM day programs allow repeating the same cycle every day of the week, weekdays only, weekends only, or any selected days.
To assure accuracy an individually serial-numbered Traceable Certificate is provided from our ISO 17025 calibration laboratory accredited by A2LA. It indicates traceability to standards provided by NIST (National Institute of Standards and Technology).