Learn More
Electron Microscopy Sciences Mini Tool Set B
SDP
Catalog No. 50996719
Encompass
Contains eight tips with integrated, color-coded handles. Length is 127 mm. Tip is 0.8mm and made from hardened tool steel, nickel plated. Contains: Straight Prober, 30 deg Prober, 90 deg Prober, Chisel, Knife, Graver, Diamond Scriber.
Catalog No. 50-996-719 Supplier Electron Microscopy Sciences Supplier No. 6209720
Contains eight tips with integrated, color-coded handles. Length is 127 mm. Tip is 0.8mm and made from hardened tool steel, nickel plated. Contains: Straight Prober, 30 deg Prober, 90 deg Prober, Chisel, Knife, Graver, Diamond Scriber.