Learn More
Electron Microscopy Sciences Mini Tool Set C
SDP
Catalog No. 50996720
Encompass
Straight Scriber with sharp tip, Sharp Triangular Scraper, Bent Prober with sharp tip, integrated hexagonal aluminum handles. Tip material is hardened steel, nickel plated. Tip size is 0.8mm by 25mm long.
Catalog No. 50-996-720 Supplier Electron Microscopy Sciences Supplier No. 6209730
Straight Scriber with sharp tip, Sharp Triangular Scraper, Bent Prober with sharp tip, integrated hexagonal aluminum handles. Tip material is hardened steel, nickel plated. Tip size is 0.8mm by 25mm long.