Learn More
Bio Rad Laboratories MINI TRANS-BLOT MODULE
Catalog No. 509010675
Encompass
Module; Bio-Rad; Mini Trans-Blot; Includes 2 gel holder cassettes, 4 fiber pads, modular electrode assembly, Bio-Ice cooling unit, instructions (without lower buffer tank and lid)
Catalog No. 50-901-0675 Supplier Bio Rad Laboratories Supplier No. 1703935
Module; Bio-Rad; Mini Trans-Blot; Includes 2 gel holder cassettes, 4 fiber pads, modular electrode assembly, Bio-Ice cooling unit, instructions (without lower buffer tank and lid)