Learn More
Research Products International Corp Mini Incubation Tray for Immunostaining 8 x 8 x 2 1/4 Inches
SDP
Catalog No. 50476555
Encompass
Specially designed tray for microwave enhanced immunostaining procedures. Built-in dividers provide channels that distribute water evenly under the slides. The water absorbs microwave energy and prevents overheating of samples. The technique allows a standard three step staining procedure to be performed in as little as fifteen minutes.
Catalog No. 50-476-555 Supplier Research Products International Corp Supplier No. 248272
Specially designed tray for microwave enhanced immunostaining procedures. Built-in dividers provide channels that distribute water evenly under the slides. The water absorbs microwave energy and prevents overheating of samples. The technique allows a standard three step staining procedure to be performed in as little as fifteen minutes.