Learn More
Bellco Glass, Inc. Mini Tube Roller Complete, 230V
SDP
Catalog No. 501945898
Encompass_Preferred
The Bellco Mini Tube Roller is a variable speed roller unit for general laboratory usage. The compact size is ideal for experimental and pilot-scale operations. Methods developed on this unit can easily be scaled up. Robustly constructed and designed for easy cleaning, with EPDM rubber rollers to assure safe positive rotating motion. The unit has 11 rollers, five of which can be removed to accommodate various bottle dimensions. When using hybridization tubes, an o-ring (A12-1067) must be fitted on the lower end to equalize the diameter of the screw cap. When alternate rollers are removed, 3 roller bottles up to 120 mm can rotate at a maximum speed of 4 rpm. MADE IN THE USA
Catalog No. 50-194-5898 Supplier Bellco Glass, Inc. Supplier No. 762230230
The Bellco Mini Tube Roller is a variable speed roller unit for general laboratory usage. The compact size is ideal for experimental and pilot-scale operations. Methods developed on this unit can easily be scaled up. Robustly constructed and designed for easy cleaning, with EPDM rubber rollers to assure safe positive rotating motion. The unit has 11 rollers, five of which can be removed to accommodate various bottle dimensions. When using hybridization tubes, an o-ring (A12-1067) must be fitted on the lower end to equalize the diameter of the screw cap. When alternate rollers are removed, 3 roller bottles up to 120 mm can rotate at a maximum speed of 4 rpm. MADE IN THE USA