Learn More
GRANT INSTRUMENTS Mini-rotator, vertical 360°, 5 to 30 rpm, holds 22 x 1.5-15ml tubes
SDP
Catalog No. NC2107387
Encompass_Preferred
PTR-25 Mini-rotator provides vertical rotation of the platform.It is an ideal instrument for preventing blood coagulations, for extraction, diffusion and dialyzing biological liquids in test tubes. PTR-25 is a compact device suitable for convenient use in small laboratories.Compact, economy solution with simple to use controls, providing thorough mixing of samples in tubes up to 50 ml. Suitable for use in cold rooms and incubators.360° vertical rotation.Vertical rotation speed range: 5 to 30 rpm.Timer with audible alarm and automatic switch-off.Very easy to operate with simple controls and easy to view LED screen.Reliable and extremely quiet motor produces regulated rotation throughout the speed range.Compact with a low profile and small footprint – fits neatly into the workspace.Platform accommodating 22 tubes (up to 15mm diameter) is included as standard. See Accessories page for additional accessories.
This item is not returnable. View return policy
Catalog No. NC2107387 Supplier GRANT INSTRUMENTS Supplier No. PTR25
PTR-25 Mini-rotator provides vertical rotation of the platform.It is an ideal instrument for preventing blood coagulations, for extraction, diffusion and dialyzing biological liquids in test tubes. PTR-25 is a compact device suitable for convenient use in small laboratories.Compact, economy solution with simple to use controls, providing thorough mixing of samples in tubes up to 50 ml. Suitable for use in cold rooms and incubators.360° vertical rotation.Vertical rotation speed range: 5 to 30 rpm.Timer with audible alarm and automatic switch-off.Very easy to operate with simple controls and easy to view LED screen.Reliable and extremely quiet motor produces regulated rotation throughout the speed range.Compact with a low profile and small footprint – fits neatly into the workspace.Platform accommodating 22 tubes (up to 15mm diameter) is included as standard. See Accessories page for additional accessories.