Learn More
Sklar Instruments Mini Tischler Universal Style Biopsy Punch, Sklar EA.
SDP
Catalog No. 501183576 Shop All Sklar Instruments Products
Encompass_Preferred
Mini Tischler Universal Style Biopsy Punch, Sklar, Sklar's Mini Tischler Universal Style Biopsy Punch Forceps are a non-ratcheted, double spring OB/GYN instrument most commonly used in colposcopy procedures to obtain cervical tissue samples for biopsy. The mini Tischler bite, 2.3mm x 4.2mm, is slightly smaller than a standard Tischler bite and takes a smaller tissue sample. The mini or baby style biopsy punch is often preferred because there is typically less bleeding and pain when the smaller tissue sample is removed. It also features a single, lower tooth used to grasp difficult to reach tissue. This product is a complete instrument which consists of a handle and a detachable cannula/tip so other universal tip styles may be used with one handle. The length of the cannula/tip is approximately 8 inches.
Catalog No. 50-118-3576 Supplier Sklar Instruments Supplier No. 907553
Mini Tischler Universal Style Biopsy Punch, Sklar, Sklar's Mini Tischler Universal Style Biopsy Punch Forceps are a non-ratcheted, double spring OB/GYN instrument most commonly used in colposcopy procedures to obtain cervical tissue samples for biopsy. The mini Tischler bite, 2.3mm x 4.2mm, is slightly smaller than a standard Tischler bite and takes a smaller tissue sample. The mini or baby style biopsy punch is often preferred because there is typically less bleeding and pain when the smaller tissue sample is removed. It also features a single, lower tooth used to grasp difficult to reach tissue. This product is a complete instrument which consists of a handle and a detachable cannula/tip so other universal tip styles may be used with one handle. The length of the cannula/tip is approximately 8 inches.