Description
Includes
Mini USB Cable
- This cable is included with the following interfaces:
- LabQuest 2
- LabQuest Stream™
- LabQuest Mini
- Original LabQuest (no longer sold)
Warranty and Services
5-year limited warranty
Specifications
Specifications
| For Use With (Equipment) | Emissions Spectrometer, Spectrophotometer, Mini GC Plus, LabQuest 2, LabQuest Stream |
| For Use With (Application) | Educational use only |
| Includes | USB mini-B plug and USB standard-A plug |
| Product Type | Replacement Part |