Learn More
Research Products International Corp Mini UV Viewing System, Includes Lamps (254/365 nm) and Cabinet
SDP
Catalog No. 50212694
Encompass
Mini UV viewing cabinet and lamp provides self contained illumination system. Multi-band shortwave/longwave (254/365nm) hand held 6 watt lamp is supplied allowing various applications. Cabinet features removable bottom panel, metal base and plastic top. Lightweight access curtain blocks out external light for sample viewing.
Catalog No. 50-212-694 Supplier Research Products International Corp Supplier No. 353060
Mini UV viewing cabinet and lamp provides self contained illumination system. Multi-band shortwave/longwave (254/365nm) hand held 6 watt lamp is supplied allowing various applications. Cabinet features removable bottom panel, metal base and plastic top. Lightweight access curtain blocks out external light for sample viewing.