Learn More
Electron Microscopy Sciences Mini Vacuum Desiccator
SDP
Catalog No. 501899543
Encompass_Preferred
The top part of the desiccator is clear, molded from polycarbonate and see-through from any angle. A polyethylene stopcock with a "PTFE" plug allows for gentle releasing of the vacuum. A peripheral flange around the polypropylene bottom section fits into and snap locks therefore sealing the two halves against a "O" ring. Vacuum retention rating is 740 mm Hg (29"). Inside dimensions are 102 mm (4) diameter x 59 mm (2 7/16)high. The base is 175 mm (6 7/8) diameter.
Catalog No. 50-189-9543 Supplier Electron Microscopy Sciences Supplier No. 7124601
The top part of the desiccator is clear, molded from polycarbonate and see-through from any angle. A polyethylene stopcock with a "PTFE" plug allows for gentle releasing of the vacuum. A peripheral flange around the polypropylene bottom section fits into and snap locks therefore sealing the two halves against a "O" ring. Vacuum retention rating is 740 mm Hg (29"). Inside dimensions are 102 mm (4) diameter x 59 mm (2 7/16)high. The base is 175 mm (6 7/8) diameter.