Learn More
Cole-Parmer Miniature Check Valve, Nylon Housing, Silicone Diaphragm, 1/4" ID
Catalog No. 502557989 Shop All Cole-Parmer Products
Encompass_Preferred
Install these miniature check valves with hose barbs in any position to prevent backflow. They are compatible with both liquids and gases for pressure or vacuum applications. They require 0.18 psi breaking pressure; closing pressure is 0.014 psi.
Catalog No. 50-255-7989 Supplier Cole-Parmer Supplier No. 5011519
Install these miniature check valves with hose barbs in any position to prevent backflow. They are compatible with both liquids and gases for pressure or vacuum applications. They require 0.18 psi breaking pressure; closing pressure is 0.014 psi.