Learn More
Alkali Scientific Mini Vertical Rack with 3″ Mini Boxes & Cell Dividers – 6 Boxes High
SDP
Catalog No. 5000663038
Encompass_Preferred
For 3″ Mini Boxes Vertical freezer racks for laboratory chest and liquid nitrogen freezers provide storage of 3” mini boxes and a total of 6 boxes per rack. These vertical freezer racks are made from durable stainless steel material. Each rack includes handles which allow for easy retrieval from the freezer. These racks also come standard with a locking rod for added security. The freezer rack comes standard with 6 storage boxes and cell dividers. Dimensions H x Wx D: 19 3/8 x 3 3/8 x 3 1/4 inch (492 x 86 x 82mm) Compartment Dimensions H x W x D: 3 1/8 x 3 1/16 x 3 1/16 inch (80 x 78 x 78mm) **Lock device is ordered separately.
Catalog No. 50-006-63038 Supplier Alkali Scientific Supplier No. MVR63
For 3″ Mini Boxes Vertical freezer racks for laboratory chest and liquid nitrogen freezers provide storage of 3” mini boxes and a total of 6 boxes per rack. These vertical freezer racks are made from durable stainless steel material. Each rack includes handles which allow for easy retrieval from the freezer. These racks also come standard with a locking rod for added security. The freezer rack comes standard with 6 storage boxes and cell dividers. Dimensions H x Wx D: 19 3/8 x 3 3/8 x 3 1/4 inch (492 x 86 x 82mm) Compartment Dimensions H x W x D: 3 1/8 x 3 1/16 x 3 1/16 inch (80 x 78 x 78mm) **Lock device is ordered separately.