Learn More
Alkali Scientific Alkali Scientific™ Mini Vertical Rack for 3″ Mini Boxes – Rack Only – 10 Boxes High
SDP
Catalog No. 503029256
Encompass_Preferred
For 3″ Mini Boxes Vertical freezer racks for laboratory chest and liquid nitrogen freezers provide storage of 3” mini boxes and a total of 10 boxes per rack. These vertical freezer racks are made from durable stainless-steel material. Each rack includes handles which allow for easy retrieval from the freezer. These racks also come standard with a locking rod for added security. The freezer rack does not come standard with storage boxes. Dimensions H x Wx D: 32 1/16 x 3 3/8 x 3 1/4 inch (814 x 86 x 82mm)Compartment Dimensions H x W x D: 3 1/8 x 3 1/16 x 3 1/16 inch (80 x 78 x 78mm) **Lock device is ordered separately.
Catalog No. 50-302-9256 Supplier Alkali Scientific Supplier No. MVR103
For 3″ Mini Boxes Vertical freezer racks for laboratory chest and liquid nitrogen freezers provide storage of 3” mini boxes and a total of 10 boxes per rack. These vertical freezer racks are made from durable stainless-steel material. Each rack includes handles which allow for easy retrieval from the freezer. These racks also come standard with a locking rod for added security. The freezer rack does not come standard with storage boxes. Dimensions H x Wx D: 32 1/16 x 3 3/8 x 3 1/4 inch (814 x 86 x 82mm)Compartment Dimensions H x W x D: 3 1/8 x 3 1/16 x 3 1/16 inch (80 x 78 x 78mm) **Lock device is ordered separately.