Learn More
BURKLE INC Mini ViscoSampler, V4A, 23.62 in. (60 cm), volume30 ml, Ø 0.59 in. (15 mm)
Supplier:  BURKLE INC 53411060
Encompass_Preferred
Sampler for viscous substances such as pastes, mud, creams, oils. The sample is drawn in using the pump rod and directly conveyed to a sampling container. Easy cleaning for flawless sampling.
Catalog No. 50-000-25778