Learn More
BURKLE INC Mini ViscoSampler, V4A, 59.06 in. (150 cm), volume75 ml, Ø 0.59 in. (15 mm)
Catalog No. 5000026074
Encompass_Preferred
Sampler for viscous substances such as pastes, mud, creams, oils. The sample is drawn in using the pump rod and directly conveyed to a sampling container. Easy cleaning for flawless sampling.
Catalog No. 50-000-26074 Supplier BURKLE INC Supplier No. 53411150
Sampler for viscous substances such as pastes, mud, creams, oils. The sample is drawn in using the pump rod and directly conveyed to a sampling container. Easy cleaning for flawless sampling.