Learn More
JDM LAB SOLUTIONS LLC MINI VORTEX MIXER 3,000 BLUE
Catalog No. NC1917955
Encompass
The Mini vortex mixer is the ideal vortex mixer for every lab with its compact size, ease of use, and its economic price point. The 3,000 rpm fixed speed ensures there is no sacrificing any mixing power. Extremely compact and easy operation. Powerful vortexing for tubes up to 30mm. Head is made of durable material to ensure long life. Refined steel base is designed to ensure product stability.
Catalog No. NC1917955 Supplier JDM LAB SOLUTIONS LLC Supplier No. VM03BU
The Mini vortex mixer is the ideal vortex mixer for every lab with its compact size, ease of use, and its economic price point. The 3,000 rpm fixed speed ensures there is no sacrificing any mixing power. Extremely compact and easy operation. Powerful vortexing for tubes up to 30mm. Head is made of durable material to ensure long life. Refined steel base is designed to ensure product stability.