Learn More
Research Products International Corp Mini Vortexer, Gray/BL
SDP
Catalog No. 502229794
Encompass
Mini Vortexer offers optimal vortex mixing for a variety of tube sizes up to 50ml. Simple, touch start operation enables quick mixing and rapid vortexing (4,500 rpm) with one hand. Modern design with stable platform features low activation force that helps prevent user fatigue when performing repetitive procedures; quiet operation; and low vibration. Maintenance free unit is manufactured of durable ABS for excellent chemical resistance and durability. Product includes Mini Vortexer, 2 Silicone Caps and Low Voltage, Double Insulated Power Adapter (100-240 VAC, 50/60 Hz) with 4 Interchangeable Plugs.
Powerful mixing
Low profile, compact size
5 year warranty
Catalog No. 50-222-9794 Supplier Research Products International Corp Supplier No. 353136
Mini Vortexer offers optimal vortex mixing for a variety of tube sizes up to 50ml. Simple, touch start operation enables quick mixing and rapid vortexing (4,500 rpm) with one hand. Modern design with stable platform features low activation force that helps prevent user fatigue when performing repetitive procedures; quiet operation; and low vibration. Maintenance free unit is manufactured of durable ABS for excellent chemical resistance and durability. Product includes Mini Vortexer, 2 Silicone Caps and Low Voltage, Double Insulated Power Adapter (100-240 VAC, 50/60 Hz) with 4 Interchangeable Plugs.
Powerful mixing
Low profile, compact size
5 year warranty