Learn More
Electron Microscopy Sciences Mini Vortexer, Gray/Purple
SDP
Catalog No. 501930744
Encompass_Preferred
This new Mini Vortexer offers immediate vortex shaking for a variety of tube sizes 2 to 50 ml. It has a simple, touch start operation which enables exceptional mixing performance using one hand.
The mixer is crafted from durable ABS and silicone materials to resist most commonly used chemicals. Save valuable space on a bench top and in fume hoods with the low profile design and small footprint. Save time with the rapid vortex action. The modern design allows easy one handed touch start operation and shaking stops when the tube is lifted. Non-slip feet on the lab vortex mixer ensures the unit stays in place while mixing and provides stability. Low activation force helps prevents user fatigue in repetitive procedures. Quiet operation and low vibration on the vortex machine helps users stay focused.
Color: Gray/Purple
Catalog No. 50-193-0744 Supplier Electron Microscopy Sciences Supplier No. 8700402
This new Mini Vortexer offers immediate vortex shaking for a variety of tube sizes 2 to 50 ml. It has a simple, touch start operation which enables exceptional mixing performance using one hand.
The mixer is crafted from durable ABS and silicone materials to resist most commonly used chemicals. Save valuable space on a bench top and in fume hoods with the low profile design and small footprint. Save time with the rapid vortex action. The modern design allows easy one handed touch start operation and shaking stops when the tube is lifted. Non-slip feet on the lab vortex mixer ensures the unit stays in place while mixing and provides stability. Low activation force helps prevents user fatigue in repetitive procedures. Quiet operation and low vibration on the vortex machine helps users stay focused.
Color: Gray/Purple