Learn More
Omnimed Inc Mini Wall Storage Cabinet W Combo Lock (12’H by 9”W by 4”D)
SDP
Catalog No. NC3956571
Encompass_Preferred
Are you short on space in your restaurant, lab, or doctor’s office? Introducing the Mini Wall Storage Cabinet W Combo Lock by Omnimed! This must-have cabinet is perfect for adding overhead storage in any office environment.Constructed from durable 22 gauge steel with a powder coat finish that will last for years to come, this wall storage cabinet with the lock will keep all of your materials protected and secure. As an added bonus, the cabinet comes with a push button combination lock, so you never have to worry about security. The cabinet measures twelve inches high by nine inches wide by four inches deep, with internal measurements of 10.25 inches high by 8.75 inches wide by 3.25 inches deep
Catalog No. NC3956571 Supplier Omnimed Inc Supplier No. 291609COMB
Are you short on space in your restaurant, lab, or doctor’s office? Introducing the Mini Wall Storage Cabinet W Combo Lock by Omnimed! This must-have cabinet is perfect for adding overhead storage in any office environment.Constructed from durable 22 gauge steel with a powder coat finish that will last for years to come, this wall storage cabinet with the lock will keep all of your materials protected and secure. As an added bonus, the cabinet comes with a push button combination lock, so you never have to worry about security. The cabinet measures twelve inches high by nine inches wide by four inches deep, with internal measurements of 10.25 inches high by 8.75 inches wide by 3.25 inches deep