Learn More
Research Products International Corp Mini Water Bath, 6L
SDP
Catalog No. 501977005
Encompass
Ideal size water bath for many laboratory applications. Compact styling enables the unit to be placed in any convenient location. A 550 watt heating element provides rapid heating and is accurately controlled with a hydraulic thermostat, additional features include: stainless steel chamber, gabled stainless steel cover, analog heat control and magnetic thermometer clip.
Catalog No. 50-197-7005 Supplier Research Products International Corp Supplier No. 246640
Ideal size water bath for many laboratory applications. Compact styling enables the unit to be placed in any convenient location. A 550 watt heating element provides rapid heating and is accurately controlled with a hydraulic thermostat, additional features include: stainless steel chamber, gabled stainless steel cover, analog heat control and magnetic thermometer clip.