Learn More
JDM LAB SOLUTIONS LLC Mini Water Baths WB-02A
Catalog No. NC2385929
Encompass
Microprocessor PID control with digital displayExcellent temperature uniformityOverheating alarm and automatic shut offDry run protection with audible alarm to avoid damage if the water level is too lowIndicator light illuminates when temperature >60°CVarious tube holders are available to order separately
Catalog No. NC2385929 Supplier JDM LAB SOLUTIONS LLC Supplier No. WB02A
Microprocessor PID control with digital displayExcellent temperature uniformityOverheating alarm and automatic shut offDry run protection with audible alarm to avoid damage if the water level is too lowIndicator light illuminates when temperature >60°CVarious tube holders are available to order separately