Learn More
Hoefer, Inc Hoefer™ Mini Transfer Tank Combination Package
SDP
Catalog No. NC2740645
Encompass_Preferred
Save time and money with a combination packageIncludes:• SE260-10A-.75 Mighty Small II Mini Deluxe Vertical Unit• TE22 Mighty Small Mini Transfer Tank• PS300B 300 V, 500 mA, 90 W Power Supply
Catalog No. NC2740645 Supplier Hoefer, Inc Supplier No. 326223
Save time and money with a combination packageIncludes:• SE260-10A-.75 Mighty Small II Mini Deluxe Vertical Unit• TE22 Mighty Small Mini Transfer Tank• PS300B 300 V, 500 mA, 90 W Power Supply