Learn More
Pyramex™ Mini Ztek™ Safety Eyewear
Catalog No. 19155546 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Lens Tint:
Amber
Blue
Blue, Mirror
Clear
Gray
Indoor/Outdoor, Mirror
Pink
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint
19-155-546 Indoor/Outdoor, Mirror
19-155-520 Clear
19-155-526 Pink
19-155-531 Gray
19-155-534 Amber
19-155-544 Blue, Mirror
19-155-539 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 19-155-546 Supplier Pyramex™ Supplier No. S2580SN
Add to Cart
Add to Cart
This item is not returnable. View return policy
Has all the great features of Ztek™ in a design better suited for slimmer facial sizes
  • Integrated nose piece
  • Soft, non-slip rubber temple tips provide non-binding fit
  • Economical wrap-around single lens provides full panoramic view
  • Scratch resistant polycarbonate lens provides 99% UVA/B/C protection
  • Specifications
    Lens Coating Anti-fog
    Certifications/Compliance ANSI Z87.1, CSA Z94.3-07
    Certifications/Compliance (Europe) EN-166
    Lens Tint Indoor/Outdoor, Mirror
    Product Type Eye and Face Protection Spectacles
    Weight (Metric) 20 g
    Material Polycarbonate
    Disposable Reusable
    Lens Material Polycarbonate
    UV Protection U6
    Frame Type Frameless