Learn More
Braintree Scientific MiniArco Professional Cordless Trimmer w/#30 blade & charger
SDP
Supplier:  Braintree Scientific CLP8787450A
Encompass
The MiniARCO Unsurpassed rotary motor is very fast and powerful Cordless operation for convenience Lightweight, quiet and cool running Superior quick detachable blade set Includes: Trimmer, charger, detachable #30 blade set, cleaning brush, blade oil, two dual-sided guide combs and instruction book Product Specifications: Cord type: Cord / Cordless Trimmer speed: Sinlge speed Unit size (hxwxd): 5.5 inches Unit weight: 5 oz. Charging time: 8 Hours Running time: 45 Minutes Strokes per minute: 6,000 Battery type: NiMH Motor type: Direct Drive Rotary Warranty: 1 Year limited warranty **Sold for research purposes only and not for human use**
Catalog No. 50-195-4535