Learn More
Fine Science Tools MINIATURE FILE SET
SDP
Catalog No. NC9391827
Encompass
Product: Miniature File Set/ Alloy/Material: Carbon Steel/ Autoclave Safe: No/ Features: 8 different file profiles included/ Total Length: 10 cm/ Unit: Pack of 8
Catalog No. NC9391827 Supplier Fine Science Tools Supplier No. 3007614
Product: Miniature File Set/ Alloy/Material: Carbon Steel/ Autoclave Safe: No/ Features: 8 different file profiles included/ Total Length: 10 cm/ Unit: Pack of 8