Learn More
Electron Microscopy Sciences Miniature Measuring Scale #2
SDP
Catalog No. 50996796
Encompass_Preferred
These unit measuring scales may be used directly under a microscope or magnifier for quick measuring of small specimens, parts, grooves, slots, line widths, holes, chambers, etc. Precision fabricated from stainless steel, the scales are welded to bendable stainless steel shafts and mounted in color coded, non-rolling handles. Each scale is individually packed in a plastic tube. Overall length of the scale is 83 mm (3 1/4"), including handle. Calibrated to 0.005", 0.2" total
Catalog No. 50-996-796 Supplier Electron Microscopy Sciences Supplier No. 6213502
These unit measuring scales may be used directly under a microscope or magnifier for quick measuring of small specimens, parts, grooves, slots, line widths, holes, chambers, etc. Precision fabricated from stainless steel, the scales are welded to bendable stainless steel shafts and mounted in color coded, non-rolling handles. Each scale is individually packed in a plastic tube. Overall length of the scale is 83 mm (3 1/4"), including handle. Calibrated to 0.005", 0.2" total