Learn More
Chemglass Life Sciences Miniature Over-Head Stirrer with Remote Digital Speed Control, Speed: 20-800 rpm, 91 in-oz Maximum Torque
SDP
Catalog No. 50401980
Encompass_Preferred
Miniature Over-Head Stirrer with Remote Digital Speed Control, Speed: 10-800 rpm, 91 in-oz Maximum Torque.More power than stirrers ten times its size!Incredible 91in-oz of power yet fits in the palm of your hand!Safety - Low Voltage - SparklessSealed motor housing isolates motor from organic vaporsLow voltage operation - Sparkless motor runs on 16 voltsConvenience - Weighs less than 1lbCE MarkedThe light weight of J-KEM's stirrers makes it easy to assemble and align the stirrer with the reaction flask. Stirrer coupling fits standard 10mm OD stirring shafts. Side mount clamp on digital speed controller.
Catalog No. 50-401-980 Supplier Chemglass Life Sciences Supplier No. CG2039A10
Miniature Over-Head Stirrer with Remote Digital Speed Control, Speed: 10-800 rpm, 91 in-oz Maximum Torque.More power than stirrers ten times its size!Incredible 91in-oz of power yet fits in the palm of your hand!Safety - Low Voltage - SparklessSealed motor housing isolates motor from organic vaporsLow voltage operation - Sparkless motor runs on 16 voltsConvenience - Weighs less than 1lbCE MarkedThe light weight of J-KEM's stirrers makes it easy to assemble and align the stirrer with the reaction flask. Stirrer coupling fits standard 10mm OD stirring shafts. Side mount clamp on digital speed controller.