Learn More
MilliporeSigma™ Supelco™ Miniature Regulator with Gauge
Catalog No. 111004735 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Miniature Regulator with Gauge
Catalog No.
11-100-4735 Miniature Regulator with Gauge
1 options
Catalog No. 11-100-4735 Supplier MilliporeSigma™ Supelco™ Supplier No. 513010
Add to Cart
Add to Cart
Reliable pressure regulation to 1 psig (0.07 kg/cm2), indicated on a 0-60 psi (4.2 kg/cm2) gauge. Easily connects to SCOTTY 14 cylinders with the 1/8-inch NPT connector provided. You can also attach the syringe adapter (Cat. No. 609010) to the regulator for low pressure sample removal. Aluminum body with acetyl resin bonnet. Maximum inlet pressure: 400 psi (28.1 kg/cm2).
TRUSTED_SUSTAINABILITY
Specifications
Description Do NOT use with Oxygen.