Learn More
Futek Advanced Sensor Tech MINIATURE S BEAM JR LOAD CELL
Catalog No. NC3780996
Encompass
LSB200 , 50 g , S-Beam Jr.® Load Cell , Material - 2024-T4 , M3x0.5-Thread , Overload Protection , Vacuum Compatible/Low Outgassing , 29 Awg 4 Conductor Spiral Shielded Teflon , 5 ft Long
Catalog No. NC3780996 Supplier Futek Advanced Sensor Tech Supplier No. QSH02409
LSB200 , 50 g , S-Beam Jr.® Load Cell , Material - 2024-T4 , M3x0.5-Thread , Overload Protection , Vacuum Compatible/Low Outgassing , 29 Awg 4 Conductor Spiral Shielded Teflon , 5 ft Long